missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-fluoropyridine, 97%, Thermo Scientific Chemicals

Catalog Number 11304406
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
21717-96-4
C5H5FN2
112.11
MFCD01861120
YJTXQLYMECWULH-UHFFFAOYSA-N
2-amino-5-fluoropyridine, 5-fluoro-2-pyridinamine, 5-fluoro-2-pyridineamine, 5-fluoro-pyridin-2-ylamine, 5-fluoro-2-pyridylamine, 2-amino 5-fluoropyridine, 2-pyridinamine, 5-fluoro, 5-fluoro-2-aminopyridine, 2-amino-5-fluoro pyridine, 5-fluoropyridimine
2737701
5-fluoropyridin-2-amine
NC1=CC=C(F)C=N1
This item is not returnable. View return policy
21717-96-4
C5H5FN2
112.11
MFCD01861120
YJTXQLYMECWULH-UHFFFAOYSA-N
2-amino-5-fluoropyridine, 5-fluoro-2-pyridinamine, 5-fluoro-2-pyridineamine, 5-fluoro-pyridin-2-ylamine, 5-fluoro-2-pyridylamine, 2-amino 5-fluoropyridine, 2-pyridinamine, 5-fluoro, 5-fluoro-2-aminopyridine, 2-amino-5-fluoro pyridine, 5-fluoropyridimine
2737701
5-fluoropyridin-2-amine
NC1=CC=C(F)C=N1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 93°C to 97°C
Odor Characteristic
Quantity 250 mg
UN Number UN2811
Beilstein 385938
Formula Weight 112.11
Percent Purity 97%
Chemical Name or Material 2-Amino-5-fluoropyridine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'