Learn More
Alfa Aesar™ 2-Amino-5-fluoro-4-methylpyridine, 95%
Brand: Alfa Aesar™ H64815.06
Additional Details : CAS Number : 301222-66-2 Weight : 0.00500kg
Quantity | 5g |
---|
Chemical Identifiers
301222-66-2 | |
126.134 | |
RKMOPXJNMQWPLH-UHFFFAOYSA-N | |
22063670 | |
CC1=CC(=NC=C1F)N |
C6H7FN2 | |
MFCD06659479 | |
2-amino-5-fluoro-4-picoline, 2-amino-5-fluoro-4-methylpyridine, 5-fluoro-4-methyl-2-pyridinamine, 2-amino-4-methyl-5-fluoropyridine, pubchem1256, 5-fluoro-4-methyl-pyridin-2-amine, 2-pyridinamine,5-fluoro-4-methyl, 2-amino-4-methyl-5-fluoro pyridine, e 5-fluoro-4-methyl-2-pyridinamine, 2-pyridinamine, 5-fluoro-4-methyl-9ci | |
5-fluoro-4-methylpyridin-2-amine |
Specifications
2-Amino-5-fluoro-4-methylpyridine | |
301222-66-2 | |
MFCD06659479 | |
2-amino-5-fluoro-4-picoline, 2-amino-5-fluoro-4-methylpyridine, 5-fluoro-4-methyl-2-pyridinamine, 2-amino-4-methyl-5-fluoropyridine, pubchem1256, 5-fluoro-4-methyl-pyridin-2-amine, 2-pyridinamine,5-fluoro-4-methyl, 2-amino-4-methyl-5-fluoro pyridine, e 5-fluoro-4-methyl-2-pyridinamine, 2-pyridinamine, 5-fluoro-4-methyl-9ci | |
CC1=CC(=NC=C1F)N | |
126.134 | |
126.13 |
5g | |
C6H7FN2 | |
9694924 | |
RKMOPXJNMQWPLH-UHFFFAOYSA-N | |
5-fluoro-4-methylpyridin-2-amine | |
22063670 | |
95% |