Få mere at vide
2-Amino-5-chlorobenzophenone Oxime-d5, TRC
Artikelnummer. 30320202
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30320202 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30320202 Leverandør Toronto Research Chemicals Leverandørnr. TRCA60373250MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Amino-5-chlorobenzophenone Oxime-d5
CAS 65854-73-1
Molekylær formel C13H6D5ClN2O
Mængde 50 mg
Synonym (2-Amino-5-chlorophenyl)phenyl-methanone Oxime-d5, 5-Chloro-2-aminobenzophenone Oxime-d5, NSC 86144-d5
InChI formel InChI=1S/C13H11ClN2O/c14-10-6-7-12(15)11(8-10)13(16-17)9-4-2-1-3-5-9/h1-8,17H,15H2/b16-13-/i1D,2D,3D,4D,5D
SMIL [2H]c1c([2H])c([2H])c(\C(=N\O)\c2cc(Cl)ccc2N)c([2H])c1[2H]
IUPAC navn (NZ)-N-[(2-amino-5-chlorophenyl)-(2,3,4,5,6-pentadeuteriophenyl)methylidene]hydroxylamine
Molekylvægt (g/mol) 251.72
Formel vægt 251.0874