Learn More
Alfa Aesar™ 2-Amino-5-chloro-6-methylpyridine, 97%
Brand: Alfa Aesar™ H58501.06
Additional Details : CAS Number : 36936-23-9 Weight : 0.00500kg
Quantity | 5g |
---|
Chemical Identifiers
36936-23-9 | |
142.586 | |
SHIKRPPKGCWKJO-UHFFFAOYSA-N | |
12251007 | |
CC1=C(C=CC(=N1)N)Cl |
C6H7ClN2 | |
MFCD09453335 | |
2-amino-5-chloro-6-methylpyridine, 2-amino-5-chloro-6-picoline, 5-chloro-6-methyl-2-pyridinamine, 6-amino-3-chloropicoline, 2-amino-5-chloro-6-methyl pyridine, 5-chloro-6-methyl-pyridin-2-ylamine, 2-pyridinamine, 5-chloro-6-methyl, pubchem5718, 6-amino-3-chloro-2-picoline, 5-chloro-6-methyl-pyridineamine | |
5-chloro-6-methylpyridin-2-amine |
Specifications
2-Amino-5-chloro-6-methylpyridine | |
5g | |
C6H7ClN2 | |
2-amino-5-chloro-6-methylpyridine, 2-amino-5-chloro-6-picoline, 5-chloro-6-methyl-2-pyridinamine, 6-amino-3-chloropicoline, 2-amino-5-chloro-6-methyl pyridine, 5-chloro-6-methyl-pyridin-2-ylamine, 2-pyridinamine, 5-chloro-6-methyl, pubchem5718, 6-amino-3-chloro-2-picoline, 5-chloro-6-methyl-pyridineamine | |
CC1=C(C=CC(=N1)N)Cl | |
142.586 | |
142.59 |
70°C to 75°C | |
36936-23-9 | |
MFCD09453335 | |
SHIKRPPKGCWKJO-UHFFFAOYSA-N | |
5-chloro-6-methylpyridin-2-amine | |
12251007 | |
97% |