Få mere at vide
2-Amino-5-butyl-6-methylpyrimidin-4(3H)-one, TRC
Artikelnummer. 30180254
Skift visning
Mængde:
250 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30180254 1 g
30180225 500 mg
30180107 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30180254 Leverandør Toronto Research Chemicals Leverandørnr. TRCA5957801G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 4038-64-6
Molekylær formel C9 H15 N3 O
Molekylvægt (g/mol) 181.23
Synonym 2-Amino-5-butyl-6-methyl-4(3H)-pyrimidinone, 2-Amino-5-butyl-6-methyl-1,4-dihydropyrimidin-4-one; 2-Amino-5-butyl-6-methyl-1H-pyrimidin-4-one, 2-Amino-5-butyl-6-methyl-3H-pyrimidin-4-one, 2-Amino-5-butyl-6-methyl-4-pyrimidinol, 2-Amino-5-butyl-6-methyl-pyrimidin-4-ol, 2-Amino-5-butyl-6-methylpyrimidin-4-ol, NSC 210586, R 31681;
IUPAC navn 2-amino-5-butyl-4-methyl-1H-pyrimidin-6-one
SMIL CCCCc1c(C)nc(N)nc1O
Tekniske data
Kemisk navn eller materiale 2-Amino-5-butyl-6-methylpyrimidin-4(3H)-one
Mængde 1 g
InChI formel InChI=1S/C9H15N3O/c1-3-4-5-7-6(2)11-9(10)12-8(7)13/h3-5H2,1-2H3,(H3,10,11,12,13)
Formel vægt 181.1215