Få mere at vide
2-Amino-5-bromopyridine-3,4,6-d3, CDN
Artikelnummer. 30313574
Skift visning
Mængde:
0.05 g
0.1 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30313574 0.1 g 0.1g
30316916 0.05 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30313574 Leverandør CDN Leverandørnr. CDND79880.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Amino-5-bromopyridine-3,4,6-D3
CAS 952109-91-0
Molekylær formel C5 D3 H2 Br N2
Mængde 0.1 g
Synonym 2-Pyridin-3,4,6-d3-amine, 5-bromo- (ACI), 5-Bromo-2-pyridin-3,4,6-d3-amine (ACI), 2-Amino-5-bromopyridine-3,4,6-d3, 2-Amino-5-bromopyridine-d3, 2-Amino-5-bromopyridine-3,4,6-D3
InChI formel InChI=1 S/C5H5BrN2/c6-4-1-2-5(7)8-3-4/h1-3 H,(H2,7,8)/i1D,2 D,3 D
SMIL [2 H]c1nc(N)c([2 H])c([2 H])c1Br
IUPAC navn 5-bromo-3,4,6-trideuteriopyridin-2-amine
Molekylvægt (g/mol) 176.0291
Formel vægt 174.9824 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity