missing translation for 'onlineSavingsMsg'
Få mere at vide

(2-Amino-5-bromophenyl)phenyl-methanone, TRC

Artikelnummer. 30309244
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30309244 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30309244 Leverandør Toronto Research Chemicals Leverandørnr. TRCA6019605G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale (2-Amino-5-bromophenyl)phenyl-methanone
CAS 39859-36-4
Molekylær formel C13H10BNO
Mængde 5 g
Synonym 2-Amino-5-bromo-benzophenone, 2-Benzoyl-4-bromoaniline, 5-Bromo-2-aminobenzophenone, NSC 263789
InChI formel InChI=1S/C13H10BrNO/c14-10-6-7-12(15)11(8-10)13(16)9-4-2-1-3-5-9/h1-8H,15H2
SMIL Nc1ccc(Br)cc1C(=O)c2ccccc2
IUPAC navn (2-amino-5-bromophenyl)-phenylmethanone
Molekylvægt (g/mol) 276.13
Formel vægt 274.9946
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.