missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-bromo-4-methylpyrimidine, 96%, Thermo Scientific™

Catalog Number p-7035952
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
17321-93-6
C5H6BrN3
188.03
MFCD00094469
LPQVTZJEIXYDQA-UHFFFAOYSA-N
2-amino-5-bromo-4-methylpyrimidine, 5-bromo-4-methyl-pyrimidin-2-ylamine, 2-amino-4-methyl-5-bromopyrimidine, 5-bromo-4-methylpyrimidin-2-ylamine, 5-bromo-4-methyl-2-pyrimidinamine, 5-bromo-4-methyl-2-aminopyrimidine, 5-bromo-4-methylpyrimidine-2-ylamine, 2-pyrimidinamine, 5-bromo-4-methyl, pubchem21500, acmc-1c2yt
18616196
5-bromo-4-methylpyrimidin-2-amine
CC1=NC(N)=NC=C1Br
This item is not returnable. View return policy
17321-93-6
C5H6BrN3
188.03
MFCD00094469
LPQVTZJEIXYDQA-UHFFFAOYSA-N
2-amino-5-bromo-4-methylpyrimidine, 5-bromo-4-methyl-pyrimidin-2-ylamine, 2-amino-4-methyl-5-bromopyrimidine, 5-bromo-4-methylpyrimidin-2-ylamine, 5-bromo-4-methyl-2-pyrimidinamine, 5-bromo-4-methyl-2-aminopyrimidine, 5-bromo-4-methylpyrimidine-2-ylamine, 2-pyrimidinamine, 5-bromo-4-methyl, pubchem21500, acmc-1c2yt
18616196
5-bromo-4-methylpyrimidin-2-amine
CC1=NC(N)=NC=C1Br

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 192°C to 194°C
Quantity 1 g
Formula Weight 188.03
Percent Purity 96%
Chemical Name or Material 2-Amino-5-bromo-4-methylpyrimidine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'