missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-bromo-3-nitropyridine, 97%, Thermo Scientific Chemicals

Catalog Number 11487197
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
6945-68-2
C5H4BrN3O2
218.01
MFCD00047441
QOOCOFOGYRQPPN-UHFFFAOYSA-N
2-amino-5-bromo-3-nitropyridine, 5-bromo-3-nitro-pyridin-2-ylamine, 5-bromo-3-nitro-2-pyridylamine, 2-amino-3-nitro-5-bromopyridine, 2-pyridinamine, 5-bromo-3-nitro, 5-bromo-3-nitro-2-pyridinamine, pubchem1105, maybridge1_001193, acmc-1b8cy, ksc496e6b
138878
5-bromo-3-nitropyridin-2-amine
NC1=NC=C(Br)C=C1[N+]([O-])=O
This item is not returnable. View return policy
6945-68-2
C5H4BrN3O2
218.01
MFCD00047441
QOOCOFOGYRQPPN-UHFFFAOYSA-N
2-amino-5-bromo-3-nitropyridine, 5-bromo-3-nitro-pyridin-2-ylamine, 5-bromo-3-nitro-2-pyridylamine, 2-amino-3-nitro-5-bromopyridine, 2-pyridinamine, 5-bromo-3-nitro, 5-bromo-3-nitro-2-pyridinamine, pubchem1105, maybridge1_001193, acmc-1b8cy, ksc496e6b
138878
5-bromo-3-nitropyridin-2-amine
NC1=NC=C(Br)C=C1[N+]([O-])=O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 206°C to 210°C
Quantity 1 g
Beilstein 383851
Formula Weight 218.01
Percent Purity 97%
Chemical Name or Material 2-Amino-5-bromo-3-nitropyridine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'