missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Amino-5-bromo-3-nitropyridine, 97%

Brand:  Alfa Aesar™ A14985.06

Additional Details : CAS Number : 6945-68-2 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 11497197

  • 485.00 DKK / 5g

In stock, ready to ship: 1

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

6945-68-2
218.01
QOOCOFOGYRQPPN-UHFFFAOYSA-N
138878
C1=C(C(=NC=C1Br)N)[N+](=O)[O-]
C5H4BrN3O2
MFCD00047441
2-amino-5-bromo-3-nitropyridine, 5-bromo-3-nitro-pyridin-2-ylamine, 5-bromo-3-nitro-2-pyridylamine, 2-amino-3-nitro-5-bromopyridine, 2-pyridinamine, 5-bromo-3-nitro, 5-bromo-3-nitro-2-pyridinamine, pubchem1105, maybridge1_001193, acmc-1b8cy, ksc496e6b
5-bromo-3-nitropyridin-2-amine

Specifications

2-Amino-5-bromo-3-nitropyridine
5g
C5H4BrN3O2
383851
QOOCOFOGYRQPPN-UHFFFAOYSA-N
5-bromo-3-nitropyridin-2-amine
138878
97%
206°C to 210°C
6945-68-2
MFCD00047441
2-amino-5-bromo-3-nitropyridine, 5-bromo-3-nitro-pyridin-2-ylamine, 5-bromo-3-nitro-2-pyridylamine, 2-amino-3-nitro-5-bromopyridine, 2-pyridinamine, 5-bromo-3-nitro, 5-bromo-3-nitro-2-pyridinamine, pubchem1105, maybridge1_001193, acmc-1b8cy, ksc496e6b
C1=C(C(=NC=C1Br)N)[N+](=O)[O-]
218.01
218.01
Videos
SDS Documents