Få mere at vide
(2-Amino-5,6-dichloroquinazolin-3(4H)-yl)acetic Acid Hydrochloride, Mikromol™
Artikelnummer. 30460737
Skift visning
Mængde:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30460737 4 x 25 mg
17991346 100 mg
17924076 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30460737 Leverandør Mikromol Leverandørnr. MM3250.0400254AMP
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
(2-Amino-5,6-dichloroquinazolin-3(4H)-yl)acetic Acid Hydrochloride, Mikromol
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (2-Amino-5,6-dichloroquinazolin-3(4H)-yl)acetic Acid Hydrochloride
CAS 2203514-95-6
Analyt- eller komponentnavne (2-Amino-5,6-dichloroquinazolin-3(4H)-yl)acetic Acid Hydrochloride
Forsendelsestilstand Room Temperature
Molekylær formel C10 H9 Cl2 N3 O2 . Cl H
Certificeringer/compliance ISO 17025
Synonym (2-Amino-5,6-dichloroquinazolin-3(4H)-yl)acetic Acid Hydrochloride, Anagrelide Related Compound B (USP), Anagrelide RC B, Anagrelide Related Compound B as Hydrochloride
InChI formel InChI=1S/C10H9Cl2N3O2.ClH/c11-6-1-2-7-5(9(6)12)3-15(4-8(16)17)10(13)14-7;/h1-2H,3-4H2,(H2,13,14)(H,16,17);1H
Løsningstype Neat
SMIL Cl.NC1=Nc2ccc(Cl)c(Cl)c2CN1CC(=O)O
IUPAC navn 2-(2-amino-5,6-dichloro-4H-quinazolin-3-yl)acetic acid;hydrochloride
Molekylvægt (g/mol) 310.56
Mængde 4 x 25 mg
Formel vægt 308.9839 g/mol