Få mere at vide
2-Amino-5-[(3,4,5-trimethoxyphenyl)methyl]-4(1H)-pyrimidinone, TRC
Artikelnummer. 30137369
Skift visning
Mængde:
10 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30137369 100 mg
30132557 25 mg
30008824 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30137369 Leverandør Toronto Research Chemicals Leverandørnr. TRCA630830100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 92440-76-1
Molekylær formel C14 H17 N3 O4
Molekylvægt (g/mol) 291.3
Synonym 2-Amino-5-(3,4,5-trimethoxybenzyl)pyrimidin-4-ol, Trimethoprim Imp. D (EP)
IUPAC navn 2-amino-5-[(3,4,5-trimethoxyphenyl)methyl]pyrimidin-4-ol
SMIL COc1cc(Cc2cnc(N)nc2O)cc(OC)c1OC
Tekniske data
Kemisk navn eller materiale 2-Amino-5-(3,4,5-trimethoxybenzyl)pyrimidin-4-ol
Analyt- eller komponentnavne 2-Amino-5-(3,4,5-trimethoxybenzyl)pyrimidin-4-ol
Renhedsgrad noter HPLC
Forsendelsestilstand Room Temperature
InChI formel InChI=1S/C14H17N3O4/c1-19-10-5-8(6-11(20-2)12(10)21-3)4-9-7-16-14(15)17-13(9)18/h5-7H,4H2,1-3H3,(H3,15,16,17,18)
Løsningstype Neat
Industry Type Pharmaceutical
Mængde 100 mg
Formel vægt 291.1219
Procent renhed >95%