missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-(trifluoromethyl)pyrimidine, 99%, Thermo Scientific™

Catalog Number p-7033276
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
16075-42-6
C5H4F3N3
163.10
MFCD00828624
NKOTXYPTXKUCDL-UHFFFAOYSA-N
2-amino-4-trifluoromethyl pyrimidine, 4-trifluoromethyl pyrimidin-2-amine, 2-amino-4-trifluoromethylpyrimidine, 2-pyrimidinamine, 4-trifluoromethyl, 4-trifluoromethyl-2-pyrimidinamine, 4-trifluoromethyl-pyrimidin-2-ylamine, 4-trifluoromethyl pyrimidin-2-ylamine, 4-trifluoromethyl pyrimidine-2-ylamine
595915
4-(trifluoromethyl)pyrimidin-2-amine
NC1=NC=CC(=N1)C(F)(F)F
This item is not returnable. View return policy
16075-42-6
C5H4F3N3
163.10
MFCD00828624
NKOTXYPTXKUCDL-UHFFFAOYSA-N
2-amino-4-trifluoromethyl pyrimidine, 4-trifluoromethyl pyrimidin-2-amine, 2-amino-4-trifluoromethylpyrimidine, 2-pyrimidinamine, 4-trifluoromethyl, 4-trifluoromethyl-2-pyrimidinamine, 4-trifluoromethyl-pyrimidin-2-ylamine, 4-trifluoromethyl pyrimidin-2-ylamine, 4-trifluoromethyl pyrimidine-2-ylamine
595915
4-(trifluoromethyl)pyrimidin-2-amine
NC1=NC=CC(=N1)C(F)(F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 68°C to 70°C
Quantity 1 g
Formula Weight 163.1
Percent Purity 99%
Chemical Name or Material 2-Amino-4-(trifluoromethyl)pyrimidine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'