Få mere at vide
2-Amino-4-(isopropyl-d7-amino)-6-chloro-triazine, TRC
Artikelnummer. 30318698
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30318698 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30318698 Leverandør Toronto Research Chemicals Leverandørnr. TRCA6117521MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Amino-4-(isopropyl-d7-amino)-6-chloro-triazine
CAS 1216649-31-8
Molekylær formel C6 D7 H3 Cl N5
Mængde 1 mg
Synonym Atrazine-desethyl D7 (Isopropyl D7), 6-Chloro-N2-[1-(methyl-d3)ethyl-1,2,2,2-d4]-1,3,5-triazine-2,4-diamine, 6-Chloro-N-(1,1,1,2,3,3,3-heptadeuteriopropan-2-yl)-1,3,5-triazine-2,4-diamine, D7 deethylatrazine
InChI formel InChI=1S/C6H10ClN5/c1-3(2)9-6-11-4(7)10-5(8)12-6/h3H,1-2H3,(H3,8,9,10,11,12)/i1D3,2D3,3D
SMIL [2H]C([2H])([2H])C([2H])(Nc1nc(N)nc(Cl)n1)C([2H])([2H])[2H]
IUPAC navn 6-chloro-2-N-(1,1,1,2,3,3,3-heptadeuteriopropan-2-yl)-1,3,5-triazine-2,4-diamine
Molekylvægt (g/mol) 194.6732
Formel vægt 194.1064