Få mere at vide
2-Amino-4-chloro-N-[(1R,2R)-2-(2,4-difluorophenyl)-2-hydroxy-1-methyl-3-(1H-1,2,4-triazol-1-yl)propyl]benzamide-d3, TRC
Artikelnummer. 30328098
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30328098 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30328098 Leverandør Toronto Research Chemicals Leverandørnr. TRCA60341210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Amino-4-chloro-N-[(1R,2R)-2-(2,4-difluorophenyl)-2-hydroxy-1-methyl-3-(1H-1,2,4-triazol-1-yl)propyl]benzamide-d3
CAS 1330169-87-3
Molekylær formel C19H15D3ClF2N5O2
Mængde 10 mg
Synonym [R-(R*,R*)]-2-Amino-4-chloro-N-[2-(2,4-difluorophenyl)-2-hydroxy-1-methyl-3-(1H-1,2,4-triazol-1-yl)propyl]benzamide-d3
InChI formel InChI=1S/C19H18ClF2N5O2/c1-11(26-18(28)14-4-2-12(20)6-17(14)23)19(29,8-27-10-24-9-25-27)15-5-3-13(21)7-16(15)22/h2-7,9-11,29H,8,23H2,1H3,(H,26,28)/t11-,19-/m1/s1/i1D3
SMIL [2H]C([2H])([2H])[C@@H](NC(=O)c1ccc(Cl)cc1N)[C@](O)(Cn2cncn2)c3ccc(F)cc3F
IUPAC navn 2-amino-4-chloro-N-[(2R,3R)-1,1,1-trideuterio-3-(2,4-difluorophenyl)-3-hydroxy-4-(1,2,4-triazol-1-yl)butan-2-yl]benzamide
Molekylvægt (g/mol) 424.85
Formel vægt 424.1305