Få mere at vide
2-Amino-4,6-dihydroxy-5-nitropyrimidine, TRC
Artikelnummer. 30326253
Skift visning
Mængde:
500 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30326253 1 g
30328682 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30326253 Leverandør Toronto Research Chemicals Leverandørnr. TRCA6049101G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Amino-4,6-dihydroxy-5-nitropyrimidine
CAS 80466-56-4
Molekylær formel C4 H4 N4 O4
Mængde 1 g
InChI formel InChI=1S/C4H4N4O4/c5-4-6-2(9)1(8(11)12)3(10)7-4/h(H4,5,6,7,9,10)
SMIL Nc1nc(O)c(c(O)n1)[N+](=O)[O-]
IUPAC navn 2-amino-5-nitropyrimidine-4,6-diol
Molekylvægt (g/mol) 172.1
Formel vægt 172.0233