Få mere at vide
2-Amino-4,5-bis(2-methoxyethoxy)benzoic Acid Ethyl Ester Hydrochloride, Mikromol™
Beskrivelse
2-Amino-4,5-bis(2-methoxyethoxy)benzoic Acid Ethyl Ester Hydrochloride
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 2-Amino-4,5-bis(2-methoxyethoxy)benzoic Acid Ethyl Ester Hydrochloride |
| CAS | 183322-17-0 |
| Analyt- eller komponentnavne | 2-Amino-4,5-bis(2-methoxyethoxy)benzoic Acid Ethyl Ester Hydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C15 H23 N O6 . Cl H |
| Certificeringer/compliance | ISO 17025 |
| Synonym | Benzoic acid, 2-amino-4,5-bis(2-methoxyethoxy)-, ethyl ester, hydrochloride (1:1), Benzoic acid, 2-amino-4,5-bis(2-methoxyethoxy)-, ethyl ester, hydrochloride |
| InChI formel | InChI=1S/C15H23NO6.ClH/c1-4-20-15(17)11-9-13(21-7-5-18-2)14(10-12(11)16)22-8-6-19-3;/h9-10H,4-8,16H2,1-3H3;1H |
| SMIL | Cl.CCOC(=O)c1cc(OCCOC)c(OCCOC)cc1N |
| IUPAC navn | ethyl 2-amino-4,5-bis(2-methoxyethoxy)benzoate;hydrochloride |
| Vis mere |