missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Amino-3-nitropyridine, 99%

Brand:  Alfa Aesar™ A14368.14

Additional Details : CAS Number : 4214-75-9 Weight : 0.02500kg

Quantity 25g

 View more versions of this product

Product Code. 11496947

  • 905.00 DKK / 25g

Estimated Shipment
18-05-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

4214-75-9
139.114
BPYHGTCRXDWOIQ-UHFFFAOYSA-N
77887
C1=CC(=C(N=C1)N)[N+](=O)[O-]
C5H5N3O2
MFCD00006314
2-amino-3-nitropyridine, 3-nitro-2-aminopyridine, 2-pyridinamine, 3-nitro, 3-nitropyridin-2-ylamine, 3-nitro-pyridin-2-ylamine, pyridine, 2-amino-3-nitro, 2-amino3-nitropyridine, 3-nitro-2-pyridinamine, 2-amino-3-nitro pyridine, 3-nitro-2-pyridylamine
3-nitropyridin-2-amine

Specifications

2-Amino-3-nitropyridine
25g
C5H5N3O2
124468
BPYHGTCRXDWOIQ-UHFFFAOYSA-N
3-nitropyridin-2-amine
77887
99%
165°C to 167°C
4214-75-9
MFCD00006314
2-amino-3-nitropyridine, 3-nitro-2-aminopyridine, 2-pyridinamine, 3-nitro, 3-nitropyridin-2-ylamine, 3-nitro-pyridin-2-ylamine, pyridine, 2-amino-3-nitro, 2-amino3-nitropyridine, 3-nitro-2-pyridinamine, 2-amino-3-nitro pyridine, 3-nitro-2-pyridylamine
C1=CC(=C(N=C1)N)[N+](=O)[O-]
139.114
139.11
Videos
SDS Documents