Få mere at vide
2-Amino-3-(methyl-d3)-4-methyl-3H-imidazo[4,5-f]quinoline, TRC
Artikelnummer. 30326255
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30326255 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30326255 Leverandør Toronto Research Chemicals Leverandørnr. TRCA60520210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Amino-3-(methyl-d3)-4-methyl-3H-imidazo[4,5-f]quinoline
CAS 82050-11-1
Molekylær formel C12 2H3 H9 N4
Mængde 10 mg
InChI formel InChI=1S/C12H12N4/c1-7-6-9-8(4-3-5-14-9)10-11(7)16(2)12(13)15-10/h3-6H,1-2H3,(H2,13,15)/i2D3
SMIL [2H]C([2H])([2H])n1c(N)nc2c3cccnc3cc(C)c12
IUPAC navn 4-methyl-3-(trideuteriomethyl)imidazo[4,5-f]quinolin-2-amine
Molekylvægt (g/mol) 215.27
Formel vægt 215.125