missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3-methyl-5-nitropyridine, 97%, Thermo Scientific™

Catalog Number 11329835
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
18344-51-9
C6H7N3O2
153.141
MFCD03095060
JLPYUDJJBGYXEZ-UHFFFAOYSA-N
2-amino-3-methyl-5-nitropyridine, 2-amino-5-nitro-3-picoline, 3-methyl-5-nitro-pyridin-2-ylamine, 3-methyl-5-nitro-2-pyridylamine, 2-pyridinamine,3-methyl-5-nitro, 3-methyl-5-nitro-2-pyridinamine, 3-methyl-5-nitro-pyridine-2-ylamine, 3-methyl-5-nitro-pyridin-2-amine, 2-amino-5-nitro-3-picoline 2-amino-3-methyl-5-nitropyridine, pubchem1274
2734446
3-methyl-5-nitropyridin-2-amine
CC1=CC(=CN=C1N)[N+](=O)[O-]
This item is not returnable. View return policy
18344-51-9
C6H7N3O2
153.141
MFCD03095060
JLPYUDJJBGYXEZ-UHFFFAOYSA-N
2-amino-3-methyl-5-nitropyridine, 2-amino-5-nitro-3-picoline, 3-methyl-5-nitro-pyridin-2-ylamine, 3-methyl-5-nitro-2-pyridylamine, 2-pyridinamine,3-methyl-5-nitro, 3-methyl-5-nitro-2-pyridinamine, 3-methyl-5-nitro-pyridine-2-ylamine, 3-methyl-5-nitro-pyridin-2-amine, 2-amino-5-nitro-3-picoline 2-amino-3-methyl-5-nitropyridine, pubchem1274
2734446
3-methyl-5-nitropyridin-2-amine
CC1=CC(=CN=C1N)[N+](=O)[O-]

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 258°C to 261°C
Quantity 1 g
Beilstein 126947
Formula Weight 153.14
Percent Purity 97%
Chemical Name or Material 2-Amino-3-methyl-5-nitropyridine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'