Få mere at vide
2-Amino-3-methyl-3H-imidazo[4,5-h]quinoline, TRC
Artikelnummer. 30314980
Skift visning
Mængde:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30314980 10 mg
30312539 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30314980 Leverandør Toronto Research Chemicals Leverandørnr. TRCA61675010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Amino-3-methyl-3H-imidazo[4,5-H]quinoline
CAS 147293-13-8
Molekylær formel C11 H10 N4
Mængde 10 mg
Synonym 3H-Imidazo[4,5-h]quinolin-2-amine, 3-methyl-, 3-Methyl-3H-imidazo[4,5-h]quinolin-2-amine, 2-Amino-3-methyl-3H-imidazo[4,5-h]quinoline
InChI formel InChI=1S/C11H10N4/c1-15-8-5-4-7-3-2-6-13-9(7)10(8)14-11(15)12/h2-6H,1H3,(H2,12,14)
SMIL Cn1c(N)nc2c3ncccc3ccc12
IUPAC navn 3-methylimidazo[4,5-h]quinolin-2-amine
Molekylvægt (g/mol) 198.22
Formel vægt 198.0905