Få mere at vide
2-Amino-3-methyl-3H-imidazo[4,5-f]quinoline-d3, TRC
Artikelnummer. 30322333
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30322333 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30322333 Leverandør Toronto Research Chemicals Leverandørnr. TRCA61672010MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Amino-3-(trideuteromethyl)-3H-Imidazo[4,5-F]-quinoline
CAS 82050-10-0
Renhedsgrad noter HPLC
Molekylær formel C11 2H3 H7 N4
Mængde 10 mg
Synonym 3H-Imidazo[4,5-f]quinolin-2-amine, 3-(methyl-d3)-, 3-(Methyl-d3)-3H-imidazo[4,5-f]quinolin-2-amine, 2-Amino-3-(methyl-d3)-3H-imidazo(4,5-f)quinoline, 2-Amino-3-methyl-3H-imidazo[4,5-f]quinoline-d3, 2-Amino-3-methyl-3H-imidazo[4,5-f]quinoline-D3 (methyl-D3)
InChI formel InChI=1S/C11H10N4/c1-15-9-5-4-8-7(3-2-6-13-8)10(9)14-11(15)12/h2-6H,1H3,(H2,12,14)/i1D3
SMIL [2H]C([2H])([2H])n1c(N)nc2c3cccnc3ccc12
IUPAC navn 3-(trideuteriomethyl)imidazo[4,5-f]quinolin-2-amine
Molekylvægt (g/mol) 201.24
Formel vægt 201.1094
Procent renhed >95