Få mere at vide
2-Amino-3-hydroxyphenazine, Mikromol™
Artikelnummer. 17958054
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17958054 100 mg
17977194 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17958054 Leverandør Mikromol Leverandørnr. MM2500.02
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2-Amino-3-hydroxyphenazine
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Amino-3-hydroxyphenazine
CAS 4569-77-1
Analyt- eller komponentnavne 2-Amino-3-hydroxyphenazine
Forsendelsestilstand Room Temperature
Molekylær formel C12 H9 N3 O
Certificeringer/compliance ISO 9001
Synonym 2-Amino-3-hydroxyphenazine
InChI formel InChI=1S/C12H9N3O/c13-7-5-10-11(6-12(7)16)15-9-4-2-1-3-8(9)14-10/h1-6,16H,13H2
SMIL Nc1cc2nc3ccccc3nc2cc1O
IUPAC navn 3-aminophenazin-2-ol
Molekylvægt (g/mol) 211.22
Mængde 100 mg
Formel vægt 211.0746 g/mol