Learn More
2-Amino-3-chloro-5-trifluoromethylpyridine, TRC
Product Code. 30326602
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
Product Code. Quantity Unit Size
30326602 5 g 5g
1 options
Product Code. 30326602 Supplier Toronto Research Chemicals Supplier No. TRCA5960705G
 Restricted Item
Forventet forsendelse: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-Amino-3-chloro-5-trifluoromethylpyridine
CAS 79456-26-1
Molecular Formula C6H4ClF3N2
Quantity 5 g
Synonym 5-(Trifluoromethyl)-3-chloro-2-pyridinylamine, 3-Chloro-5-(trifluoromethyl)-2-pyridinamine
InChI Formula InChI=1S/C6H4ClF3N2/c7-4-1-3(6(8,9)10)2-12-5(4)11/h1-2H,(H2,11,12)
SMILES C1=C(C=NC(=C1Cl)N)C(F)(F)F
IUPAC Name 3-chloro-5-(trifluoromethyl)pyridin-2-amine
Molecular Weight (g/mol) 196.56