Få mere at vide
2-Amino-3-bromo-5-chlorobenzoic Acid, TRC
Artikelnummer. 30327188
Skift visning
Mængde:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30327188 25 g
30325673 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30327188 Leverandør Toronto Research Chemicals Leverandørnr. TRCA60201525G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Amino-3-bromo-5-chlorobenzoic Acid
CAS 41198-02-1
Molekylær formel C7H5BrClNO2
Mængde 25 g
Synonym 2-Amino-3-bromo-5-chloro-benzoic Acid, 3-Bromo-5-chloroanthranilic Acid
InChI formel InChI=1S/C7H5BrClNO2/c8-5-2-3(9)1-4(6(5)10)7(11)12/h1-2H,10H2,(H,11,12)
SMIL Nc1c(Br)cc(Cl)cc1C(=O)O
IUPAC navn 2-amino-3-bromo-5-chlorobenzoic acid
Molekylvægt (g/mol) 250.48
Formel vægt 248.9192