Få mere at vide
2-Amino-3-benzoyl-Alpha-oxo-benzeneacetic Acid Sodium Salt, TRC
Artikelnummer. 30004590
Skift visning
Mængde:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30004590 5 mg
30003573 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30004590 Leverandør Toronto Research Chemicals Leverandørnr. TRCA5937105MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 126849-31-8
Molekylær formel C15H10NNaO4
Molekylvægt (g/mol) 291.23
Synonym 2-Amino-3-benzoyl-α-oxo-benzeneacetic Acid Monosodium Salt
IUPAC navn sodium;2-(2-amino-3-benzoylphenyl)-2-oxoacetate
SMIL NC1=C(C(C(O[Na])=O)=O)C=CC=C1C(C2=CC=CC=C2)=O
Tekniske data
Kemisk navn eller materiale 2-Amino-3-benzoyl-alpha-oxo-benzeneacetic Acid Sodium Salt
Analyt- eller komponentnavne 2-Amino-3-benzoyl-alpha-oxo-benzeneacetic Acid Sodium Salt
Forsendelsestilstand Room Temperature
InChI formel InChI=1S/C15H11NO4.Na/c16-12-10(13(17)9-5-2-1-3-6-9)7-4-8-11(12)14(18)15(19)20;/h1-8H,16H2,(H,19,20);/q;+1/p-1
Løsningstype Neat
Industry Type Forensics