Få mere at vide
2-Amino-3,4,8-trimethyl-3H-imidazo[4,5-f]quinoxaline-d3, TRC
Artikelnummer. 30335368
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30335368 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30335368 Leverandør Toronto Research Chemicals Leverandørnr. TRCA63100210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Amino-3,4,8-trimethyl-3H-imidazo[4,5-f]quinoxaline-d3
CAS 1216997-87-3
Molekylær formel C12H10D3N5
Mængde 10 mg
Synonym 2-Amino-3,4,8-trimethyl-3H-imidazo[4,5-f]quinoxaline-d3, 3,4,8-Trimethyl-3H-imidazo[4,5-f]quinoxalin-2-amine-d3, 4,8-DiMeIQx-d3
InChI formel InChI=1S/C12H13N5/c1-6-4-8-9(15-7(2)5-14-8)10-11(6)17(3)12(13)16-10/h4-5H,1-3H3,(H2,13,16)/i3D3
SMIL CC1=CN=C(C=C(C)C2=C3N=C(N)N2C([2H])([2H])[2H])C3=N1
IUPAC navn 4,8-dimethyl-3-(methyl-d3)-3H-imidazo[4,5-f]quinoxalin-2-amine
Molekylvægt (g/mol) 230.28
Formel vægt 230.14