missing translation for 'onlineSavingsMsg'
Få mere at vide

2-Amino-2-norbornanecarboxylic Acid, TRC

Product Code. 30310659
Change view
Click to view available options
Mængde:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30310659 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30310659 Supplier Toronto Research Chemicals Supplier No. TRCA61905050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 2-Amino-2-norbornanecarboxylic Acid
CAS 20448-79-7
Molekylær formel C8H13NO2
Mængde 50 mg
Synonym 2-Aminobicyclo[2.2.1]heptane-2-carboxylic Acid, 2-Aminonorbornane-2-carboxylic Acid, 2-Aminonorbornyl 2-carboxylic Acid, BCH
SMIL N[C@@]1(C[C@@H]2CC[C@H]1C2)C(=O)O
Molekylvægt (g/mol) 155.19
Formel vægt 155.0946
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.