Få mere at vide
2-Amino-2-methyl-d3-propane-1,1,1,3,3,3-d6 HBr, CDN
Artikelnummer. 30317387
Skift visning
Mængde:
1 g
Artikelnummer. Mængde
30317387 1 g
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30317387 Leverandør CDN Leverandørnr. CDND68861G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1,1,1,3,3,3-hexadeuterio-2-(trideuteriomethyl)propan-2-amine hydrobromide
CAS 134071-63-9
Molekylær formel C4 2H9 H2 N . Br H
Mængde 1 g
InChI formel InChI=1 S/C4H11N.BrH/c1-4(2,3)5;/h5H2,1-3H3;1 H/i1D3,2D3,3D3;
SMIL Br.[2 H]C([2 H])([2 H])C(N)(C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H]
IUPAC navn 1,1,1,3,3,3-hexadeuterio-2-(trideuteriomethyl)propan-2-amine;hydrobromide
Molekylvægt (g/mol) 163.104
Formel vægt 162.072 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity