Få mere at vide
2-Amino-2-methyl-d3-propane-1,1,1,3,3,3-d6, CDN
Artikelnummer. 30313661
Skift visning
Mængde:
0.5 g
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30313661 0.5 g
30320838 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30313661 Leverandør CDN Leverandørnr. CDND03550.5G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale tert-Butyl-d9-amine
CAS 8-5-6045
Molekylær formel C4 D9 H2 N
Mængde 0.5 g
Synonym 2-Propan-1,1,1,3,3,3-d6-amine, 2-(methyl-d3)- (9 CI), tert-Butyl-d9-amine (7 CI, 8 CI), 1,1,1,3,3,3-Hexadeuterio-2-(trideuteriomethyl)propan-2-amine, D9-tert-Butylamine, tert-Butylamine-D9
InChI formel InChI=1 S/C4H11N/c1-4(2,3)5/h5H2,1-3H3/i1D3,2D3,3D3
SMIL [2 H]C([2 H])([2 H])C(N)(C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H]
IUPAC navn 1,1,1,3,3,3-hexadeuterio-2-(trideuteriomethyl)propan-2-amine
Molekylvægt (g/mol) 82.1923
Formel vægt 82.1456 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity