Få mere at vide
2-Amino-2-methyl-1-propanol-d11, TRC
Artikelnummer. 30323940
Skift visning
Mængde:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30323940 1 mg
30318882 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30323940 Leverandør Toronto Research Chemicals Leverandørnr. TRCA6174011MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Amino-2-methyl-1-propanol-D11
CAS 1435933-80-4
Molekylær formel C4 D11 N O
Mængde 1 mg
Synonym N,N,1,1,1,3,3,3-Octadeuterio-2-(dideuterio-deuteriooxymethyl)propan-2-amine, 2-(Amino-d2)-2-(methyl-d3)-1-propan-1,1,3,3,3-d5-ol-d, 2-Amino-2-methyl-1-propanol-d11, 2-Amino-2-methyl-1-propanol-D11
InChI formel InChI=1S/C4H11NO/c1-4(2,5)3-6/h6H,3,5H2,1-2H3/i1D3,2D3,3D2,6D/hD2
SMIL [2H]OC([2H])([2H])C(N([2H])[2H])(C([2H])([2H])[2H])C([2H])([2H])[2H]
IUPAC navn N,N,1,1,1,3,3,3-octadeuterio-2-[dideuterio(deuteriooxy)methyl]propan-2-amine
Molekylvægt (g/mol) 100.204
Formel vægt 100.1531