missing translation for 'onlineSavingsMsg'
Få mere at vide

2-Amino-1-methyl-6-phenylimidazo[4,5-b]pyridine, TRC

Product Code. 18083930
Change view
Click to view available options
Mængde:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
18083930 10 mg
18033040 25 mg
18021740 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18083930 Supplier Toronto Research Chemicals Supplier No. TRCA61700010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 105650-23-5
Molekylær formel C13 H12 N4
Molekylvægt (g/mol) 224.26
Synonym 1-Methyl-6-phenyl-1H-imidazo[4,5-b]pyridin-2-amine, PhIP;
IUPAC navn 1-methyl-6-phenylimidazo[4,5-b]pyridin-2-amine
SMIL Cn1c(N)nc2ncc(cc12)c3ccccc3

Specifications

Kemisk navn eller materiale 2-Amino-1-methyl-6-phenylimidazo[4,5-b]pyridine
Renhedsgrad noter HPLC
Mængde 10 mg
InChI formel InChI=1S/C13H12N4/c1-17-11-7-10(9-5-3-2-4-6-9)8-15-12(11)16-13(17)14/h2-8H,1H3,(H2,14,15,16)
Formel vægt 224.1062
Procent renhed >95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.