Få mere at vide
2-Amino-1-methyl-6-phenylimidazo[4,5-b]pyridine-d3, TRC
Artikelnummer. 30317465
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30317465 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30317465 Leverandør Toronto Research Chemicals Leverandørnr. TRCA6173501MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Amino-1-(trideuteromethyl)-6-Phenylimidazo[4,5-b] pyridine
CAS 210049-13-1
Renhedsgrad noter HPLC
Molekylær formel C13 2H3 H9 N4
Mængde 1 mg
InChI formel InChI=1S/C13H12N4/c1-17-11-7-10(9-5-3-2-4-6-9)8-15-12(11)16-13(17)14/h2-8H,1H3,(H2,14,15,16)/i1D3
SMIL [2H]C([2H])([2H])n1c(N)nc2ncc(cc12)c3ccccc3
IUPAC navn 6-phenyl-1-(trideuteriomethyl)imidazo[4,5-b]pyridin-2-amine
Molekylvægt (g/mol) 227.28
Formel vægt 227.125
Procent renhed >95