missing translation for 'onlineSavingsMsg'
Få mere at vide

2-Amino-1-hydroxyethane-1,1-diphosphonic Acid, TRC

Artikelnummer. 30325843
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30325843 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30325843 Leverandør Toronto Research Chemicals Leverandørnr. TRCA62798050MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 2-Amino-1-hydroxyethane-1,1-diphosphonic Acid
CAS 41003-10-5
Molekylær formel C2 H9 N O7 P2
Mængde 50 mg
Synonym P,P'-(2-amino-1-hydroxyethylidene)bisphosphonic acid, Phosphonic acid, (2-amino-1-hydroxyethylidene)bis- (9CI), 2-Amino-1-hydroxyethane-1,1-diphosphonic acid, 2-Amino-1-hydroxyethylene-1,1-bisphosphonic acid, Ethane-1-hydroxy-2-amino-1,1-diphosphonic acid, Glycine Bisphosphonate
InChI formel InChI=1S/C2H9NO7P2/c3-1-2(4,11(5,6)7)12(8,9)10/h4H,1,3H2,(H2,5,6,7)(H2,8,9,10)
SMIL NCC(O)(P(=O)(O)O)P(=O)(O)O
IUPAC navn (2-amino-1-hydroxy-1-phosphonoethyl)phosphonic acid
Molekylvægt (g/mol) 221.04
Formel vægt 220.9854
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.