Få mere at vide
2-Amino-1-(4-methylphenyl)ethanone-d3 Hydrochloride, TRC
Artikelnummer. 30323632
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30323632 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30323632 Leverandør Toronto Research Chemicals Leverandørnr. TRCA60919250MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Amino-1-(4-methylphenyl)ethanone-d3 Hydrochloride
CAS 1794977-34-6
Molekylær formel C9H9D3ClNO
Mængde 50 mg
Synonym 2-Amino-4'-methylacetophenone-d3 Hydrochloride, 4-Methylphenacylamine-d3 Hydrochloride, 2-Amino-4'-methyl-acetophenone-d3 Hydrochloride
InChI formel InChI=1S/C9H11NO.ClH/c1-7-2-4-8(5-3-7)9(11)6-10;/h2-5H,6,10H2,1H3;1H/i1D3;
SMIL Cl.[2H]C([2H])([2H])c1ccc(cc1)C(=O)CN
IUPAC navn 2-amino-1-[4-(trideuteriomethyl)phenyl]ethanone;hydrochloride
Molekylvægt (g/mol) 188.67
Formel vægt 188.0796