Få mere at vide
2-Amino-1-(4-methylphenyl)-1-propanone Hydrochloride, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | nor-Mephedrone (hydrochloride) |
| CAS | 6941-17-9 |
| Molekylær formel | C10 H13 N O . H Cl |
| Mængde | 25 mg |
| Synonym | 1-Propanone, 2-amino-1-(4-methylphenyl)-, hydrochloride (1:1), 1-Propanone, 2-amino-1-(4-methylphenyl)-, hydrochloride (9CI), Propiophenone, 2-amino-4'-methyl-, hydrochloride (6CI,7CI,8CI), 2-Amino-1-(4-methylphenyl)-1-propanone hydrochloride (1:1), NSC 60487, 2-Amino-1-(4-methylphenyl)propan-1-one hydrochloride, nor-Mephedrone hydrochloride, Mephedrone N-desmethyl metabolite hydrochloride, (±)-4-Methylcathinone hydrochloride |
| InChI formel | InChI=1S/C10H13NO.ClH/c1-7-3-5-9(6-4-7)10(12)8(2)11;/h3-6,8H,11H2,1-2H3;1H |
| SMIL | Cl.CC(N)C(=O)c1ccc(C)cc1 |
| IUPAC navn | 2-amino-1-(4-methylphenyl)propan-1-one;hydrochloride |
| Molekylvægt (g/mol) | 199.7 |
| Formel vægt | 199.0764 |