Få mere at vide
2-Amino-1,3-propanediol-1,1,2,3,3-d5, CDN
Artikelnummer. 30324460
Skift visning
Mængde:
0.05 g
0.1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30324460 0.05 g
30308954 0.1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30324460 Leverandør CDN Leverandørnr. CDND80040.05G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Amino-1,3-propanediol-1,1,2,3,3-D5
CAS 2708287-37-8
Molekylær formel C3 D5 H4 N O2
Mængde 0.05 g
Synonym 2-Amino-1,3-propanediol-1,1,2,3,3-D5, 1,3-Dihydroxy-2-propylamine-1,1,2,3,3-d5, 2-Aminopropane-1,3-diol-1,1,2,3,3-d5, 2-Hydroxy-1-(hydroxymethyl-d2)ethyl-1,2,2-d3-amine, Serinol-d5, Serinol-D5
InChI formel InChI=1 S/C3H9NO2/c4-3(1-5)2-6/h3,5-6 H,1-2,4H2/i1D2,2D2,3 D
SMIL [2 H]C([2 H])(O)C([2 H])(N)C([2 H])([2 H])O
IUPAC navn 2-amino-1,1,2,3,3-pentadeuteriopropane-1,3-diol
Molekylvægt (g/mol) 96.1399
Formel vægt 96.0947 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity