Få mere at vide
2-Amino-1-(2,5-dimethoxyphenyl)ethanol, Mikromol™
Artikelnummer. 30051282
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30051282 25 mg
17971856 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30051282 Leverandør Mikromol Leverandørnr. MM3328.010025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2-Amino-1-(2,5-dimethoxyphenyl)ethanol
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Amino-1-(2,5-dimethoxyphenyl)ethanol
CAS 3600-87-1
Analyt- eller komponentnavne 2-Amino-1-(2,5-dimethoxyphenyl)ethanol
Forsendelsestilstand Room Temperature
Molekylær formel C10 H15 N O3
Certificeringer/compliance ISO 17025
Synonym 2-Amino-1-(2,5-dimethoxyphenyl)ethanol, Midodrine Related Compound A
InChI formel InChI=1S/C10H15NO3/c1-13-7-3-4-10(14-2)8(5-7)9(12)6-11/h3-5,9,12H,6,11H2,1-2H3
SMIL COc1ccc(OC)c(c1)C(O)CN
IUPAC navn 2-amino-1-(2,5-dimethoxyphenyl)ethanol
Molekylvægt (g/mol) 197.23
Mængde 25 mg
Formel vægt 197.1052 g/mol