missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Acetylbenzoic acid, 98+%, Thermo Scientific Chemicals
217.11 DKK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
10 g
50 g
250 g
Description
2-Acetylbenzoic acid is used as a component of hair dyes. It is used as an intermediate for pharmaceutical, cosmetics, agrochemical and food industries.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Acetylbenzoic acid is used as a component of hair dyes. It is used as an intermediate for pharmaceutical, cosmetics, agrochemical and food industries.
Solubility
Soluble in Dichloromethane and Ethyl Acetate. Slightly soluble in water.
Notes
Store at -20°C. Store away from oxidizing agents. Store in a cool, dry condition in well sealed containers.
2-Acetylbenzoic acid is used as a component of hair dyes. It is used as an intermediate for pharmaceutical, cosmetics, agrochemical and food industries.
Solubility
Soluble in Dichloromethane and Ethyl Acetate. Slightly soluble in water.
Notes
Store at -20°C. Store away from oxidizing agents. Store in a cool, dry condition in well sealed containers.
Specifications
Melting Point | 115°C to 118°C |
Odor | Odorless |
Quantity | 10 g |
Beilstein | 1210557 |
Solubility Information | Soluble in Dichloromethane and Ethyl Acetate. Slightly soluble in water. |
Formula Weight | 164.16 |
Percent Purity | ≥98% |
Chemical Name or Material | 2-Acetylbenzoic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'