Få mere at vide
2-(Acetylamino)-6-[[[(1S)-1-(3-ethoxy-4-methoxyphenyl)-2-(methylsulfonyl)ethyl]amino]carbonyl]-benzoic Acid, TRC
Artikelnummer. 30136323
Skift visning
Mængde:
10 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30136323 100 mg
30133400 10 mg
30007464 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30136323 Leverandør Toronto Research Chemicals Leverandørnr. TRCA168785100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1809170-71-5
Molekylær formel C22H26N2O8S
Molekylvægt (g/mol) 478.52
Synonym Benzoic acid, 2-(acetylamino)-6-[[[(1S)-1-(3-ethoxy-4-methoxyphenyl)-2-(methylsulfonyl)ethyl]amino]carbonyl]- (ACI), 2-(Acetylamino)-6-[[[(1S)-1-(3-ethoxy-4-methoxyphenyl)-2-(methylsulfonyl)ethyl]amino]carbonyl]benzoic acid (ACI)
IUPAC navn 2-acetamido-6-[[(1S)-1-(3-ethoxy-4-methoxyphenyl)-2-methylsulfonylethyl]carbamoyl]benzoic acid
SMIL O=C(C1=C(C(O)=O)C(NC(C)=O)=CC=C1)N[C@H](CS(=O)(C)=O)C2=CC(OCC)=C(OC)C=C2
Tekniske data
Kemisk navn eller materiale 2-(Acetylamino)-6-[([(1S)-1-(3-ethoxy-4-methoxyphenyl)-2-(methylsulfonyl)ethyl]amino)carbonyl]-benzoic Acid
Analyt- eller komponentnavne 2-(Acetylamino)-6-[([(1S)-1-(3-ethoxy-4-methoxyphenyl)-2-(methylsulfonyl)ethyl]amino)carbonyl]-benzoic Acid
Forsendelsestilstand Room Temperature
InChI formel InChI=1S/C22H26N2O8S/c1-5-32-19-11-14(9-10-18(19)31-3)17(12-33(4,29)30)24-21(26)15-7-6-8-16(23-13(2)25)20(15)22(27)28/h6-11,17H,5,12H2,1-4H3,(H,23,25)(H,24,26)(H,27,28)/t17-/m1/s1
Løsningstype Neat
Industry Type Pharmaceutical
Mængde 100 mg
Formel vægt 478.141