Få mere at vide
2-Acetylamino-5-chloropyridine, Mikromol™
Artikelnummer. 30052645
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30052645 100 mg
30050546 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30052645 Leverandør Mikromol Leverandørnr. MM0149.50
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2-Acetylamino-5-chloropyridine
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Acetylamino-5-chloropyridine
CAS 45965-30-8
Analyt- eller komponentnavne 2-Acetylamino-5-chloropyridine
Forsendelsestilstand Room Temperature
Molekylær formel C7 H7 Cl N2 O
Certificeringer/compliance ISO 17025
Synonym 2-Acetylamino-5-chloropyridine
InChI formel InChI=1S/C7H7ClN2O/c1-5(11)10-7-3-2-6(8)4-9-7/h2-4H,1H3,(H,9,10,11)
SMIL CC(=O)Nc1ccc(Cl)cn1
IUPAC navn N-(5-chloropyridin-2-yl)acetamide
Molekylvægt (g/mol) 170.6
Mængde 100 mg
Formel vægt 170.0247 g/mol