missing translation for 'onlineSavingsMsg'
Learn More

2-(Acetylamino)-2-[2-(4-bromo-2-chlorophenyl)ethyl]propanedioic Acid 1,3-Diethyl Ester, TRC

Product Code. 30312120
Change view
Click to view available options
Mængde:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30312120 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30312120 Supplier Toronto Research Chemicals Supplier No. TRCA17094025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 2-(Acetylamino)-2-[2-(4-bromo-2-chlorophenyl)ethyl]propanedioic Acid 1,3-Diethyl Ester
CAS 1346604-54-3
Molekylær formel C17H21BrClNO5
Mængde 25 mg
Synonym Diethyl 2-[2-(4-Bromo-2-chlorophenyl)ethyl]-2-[(tert-butoxycarbonyl)amino]malonate
InChI formel InChI=1S/C17H21BrClNO5/c1-4-24-15(22)17(20-11(3)21,16(23)25-5-2)9-8-12-6-7-13(18)10-14(12)19/h6-7,10H,4-5,8-9H2,1-3H3,(H,20,21)
SMIL CCOC(=O)C(CCc1ccc(Br)cc1Cl)(NC(=O)C)C(=O)OCC
IUPAC navn diethyl 2-acetamido-2-[2-(4-bromo-2-chlorophenyl)ethyl]propanedioate
Molekylvægt (g/mol) 434.71
Formel vægt 433.0292
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.