missing translation for 'onlineSavingsMsg'
Få mere at vide

(±)-2-Acetoxybutyric Acid, TRC

Artikelnummer. 30310023
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
0.5 mg
Pakningsstørrelse:
0.5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30310023 0.5 mg 0.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30310023 Leverandør Toronto Research Chemicals Leverandørnr. TRCA1626650.5MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale (±)-2-Acetoxybutyric Acid
CAS 19947-43-4
Molekylær formel C6H10O4
Mængde 0.5 mg
Synonym 2-(Acetyloxy)butanoic Acid, (±)-2-(Acetyloxy)butanoic Acid, (±)-2-Acetoxybutanoic Acid, 2-Acetoxybutyric Acid, 2-Hydroxy-butyric Acid
InChI formel InChI=1S/C6H10O4/c1-3-5(6(8)9)10-4(2)7/h5H,3H2,1-2H3,(H,8,9)
SMIL CCC(OC(=O)C)C(=O)O
IUPAC navn 2-acetyloxybutanoic acid
Molekylvægt (g/mol) 146.14
Formel vægt 146.0579
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.