Få mere at vide
2-Acetoxybenzoic-3,4,5,6-d4 Acid, CDN
Artikelnummer. 30326318
Skift visning
Mængde:
0.01 g
0.05 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30326318 0.05 g
30323092 0.01 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30326318 Leverandør CDN Leverandørnr. CDND62900.05G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Acetylsalicylic Acid-d4
CAS 97781-16-3
Molekylær formel C9 2H4 H4 O4
Mængde 0.05 g
InChI formel InChI=1 S/C9H8O4/c1-6(10)13-8-5-3-2-4-7(8)9(11)12/h2-5 H,1H3,(H,11,12)/i2D,3 D,4 D,5 D
SMIL [2 H]c1c([2 H])c([2 H])c(C(=O)O)c(OC(=O)C)c1[2 H]
IUPAC navn 2-acetyloxy-3,4,5,6-tetradeuteriobenzoic acid
Molekylvægt (g/mol) 184.18
Formel vægt 184.0674 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity