Få mere at vide
2-Acetamido-6-chloro-9-(2’,3’,5’-tri-O-acetyl-Beta-D-ribofuranosyl)purine, TRC
Artikelnummer. 30328523
Skift visning
Mængde:
100 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30328523 100 mg
30320171 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30328523 Leverandør Toronto Research Chemicals Leverandørnr. TRCA149500100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Acetamido-6-chloro-9-(2',3',5'-tri-O-acetyl-beta-D-ribofuranosyl)purine
CAS 137896-02-7
Molekylær formel C18H20ClN5O8
Mængde 100 mg
Synonym N-[6-Chloro-9-(2’,3’,5’-tri-O-acetyl-beta-D-ribofuranosyl)-9H-purin-2-yl]acetamide
InChI formel InChI=1S/C18H20ClN5O8/c1-7(25)21-18-22-15(19)12-16(23-18)24(6-20-12)17-14(31-10(4)28)13(30-9(3)27)11(32-17)5-29-8(2)26/h6,11,13-14,17H,5H2,1-4H3,(H,21,22,23,25)/t11-,13-,14-,17-/m1/s1
SMIL CC(=O)Nc1nc(Cl)c2ncn([C@@H]3O[C@H](COC(=O)C)[C@@H](OC(=O)C)[C@H]3OC(=O)C)c2n1
IUPAC navn [(2R,3R,4R,5R)-5-(2-acetamido-6-chloropurin-9-yl)-3,4-diacetyloxyoxolan-2-yl]methyl acetate
Molekylvægt (g/mol) 469.83
Formel vægt 469.1