missing translation for 'onlineSavingsMsg'
Learn More

2,9-Dimethyl-1,10-phenanthroline, 98%, Thermo Scientific Chemicals

Catalog Number p-7042768
Click to view available options
Quantity:
5 g
25 g
484-11-7
C14H12N2
208.264
MFCD00004973
IYRGXJIJGHOCFS-UHFFFAOYSA-N
neocuproine, neocuproin, 1,10-phenanthroline, 2,9-dimethyl, 2,9-dimethylphenanthroline, 2,9-dimethyl-o-phenanthroline, neo-cuproin, neocuproine hemihydrate, dmphen, unii-8e7d2sh3bv, 8e7d2sh3bv
65237
2,9-dimethyl-1,10-phenanthroline
CC1=NC2=C(C=C1)C=CC3=C2N=C(C=C3)C
484-11-7
C14H12N2
208.264
MFCD00004973
IYRGXJIJGHOCFS-UHFFFAOYSA-N
neocuproine, neocuproin, 1,10-phenanthroline, 2,9-dimethyl, 2,9-dimethylphenanthroline, 2,9-dimethyl-o-phenanthroline, neo-cuproin, neocuproine hemihydrate, dmphen, unii-8e7d2sh3bv, 8e7d2sh3bv
65237
2,9-dimethyl-1,10-phenanthroline
CC1=NC2=C(C=C1)C=CC3=C2N=C(C=C3)C

Reagent for the colorimetric determination of copper

2,9-Dimethyl-1,10-phenanthroline is used as a reagent for the colorimetric determination of copper and acts as a chelating agent. It is also used in the synthesis of arly ketones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,9-Dimethyl-1,10-phenanthroline is used as a reagent for the colorimetric determination of copper and acts as a chelating agent. It is also used in the synthesis of arly ketones.

Solubility
Slightly soluble in water and light petroleum. Soluble in methanol, ethanol, acetone, ether and benzene.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 159°C to 164°C
Color Brown to White
Quantity 5 g
Sensitivity Light sensitive
Merck Index 14,6449
Solubility Information Slightly soluble in water and light petroleum; Soluble in methanol,ethanol,acetone,ether and benzene.
Formula Weight 208.26
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2,9-Dimethyl-1,10-phenanthroline

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'