Få mere at vide
2,8-Bis-(trifluoromethyl)-4-quinolinecarboxylic Acid, Mikromol™
Artikelnummer. 30052664
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30052664 25 mg
30050210 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30052664 Leverandør Mikromol Leverandørnr. MM1198.020025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2,8-Bis-(trifluoromethyl)-4-quinolinecarboxylic Acid
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Carboxymefloquine
CAS 35853-50-0
Analyt- eller komponentnavne Carboxymefloquine
Forsendelsestilstand Room Temperature
Molekylær formel C12 H5 F6 N O2
Certificeringer/compliance ISO 17025
Synonym 4-Quinolinecarboxylic acid, 2,8-bis(trifluoromethyl)-Ro 21-5104
InChI formel InChI=1S/C12H5F6NO2/c13-11(14,15)7-3-1-2-5-6(10(20)21)4-8(12(16,17)18)19-9(5)7/h1-4H,(H,20,21)
SMIL OC(=O)c1cc(nc2c(cccc12)C(F)(F)F)C(F)(F)F
IUPAC navn 2,8-bis(trifluoromethyl)quinoline-4-carboxylic acid
Molekylvægt (g/mol) 309.16
Mængde 25 mg
Formel vægt 309.0224 g/mol