missing translation for 'onlineSavingsMsg'
Få mere at vide

2,7-Dinitrofluorene, TRC

Product Code. 30385322
Change view
Click to view available options
Mængde:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30385322 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30385322 Supplier Toronto Research Chemicals Supplier No. TRCD4799251G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 2,7-Dinitrofluorene
CAS 5405-53-8
Molekylær formel C13 H8 N2 O4
Mængde 1 g
Synonym 9H-Fluorene, 2,7-dinitro- (9CI, ACI), 2,7-Dinitro-9H-fluorene (ACI), Fluorene, 2,7-dinitro- (7CI, 8CI), 2,7-Dinitrofluorene, NSC 5180
InChI formel InChI=1S/C13H8N2O4/c16-14(17)10-1-3-12-8(6-10)5-9-7-11(15(18)19)2-4-13(9)12/h1-4,6-7H,5H2
SMIL [O-][N+](=O)c1ccc2c(Cc3cc(ccc23)[N+](=O)[O-])c1
IUPAC navn 2,7-dinitro-9H-fluorene
Molekylvægt (g/mol) 256.21
Formel vægt 256.0484
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.