missing translation for 'onlineSavingsMsg'
Learn More

2,7-Dimethylpyrazolo[1,5-a]pyrimidine-6-carboxylic acid, 97%, Thermo Scientific™

Catalog Number p-2696757
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
175201-51-1
C9H9N3O2
191.19
MFCD00067901
CACYYYIWDACOAQ-UHFFFAOYSA-N
2,7-dimethylpyrazolo 1,5-a pyrimidine-6-carboxylic acid, pyrazolo 1,5-a pyrimidine-6-carboxylic acid, 2,7-dimethyl, maybridge1_004672, 6-carboxy-2,7-dimethylpyrazolo 1,5-a pyrimidine, pyrazolo 1,5-a pyrimidine-6-carboxylicacid, 2,7-dimethyl, 2,7-dimethylpyrazolo 1,5-a pyrimidine-6-carboxylicacid, 2,7-dimethyl-8-hydropyrazolo 1,5-a pyrimidine-6-carboxylic acid
2736303
2,7-dimethylpyrazolo[1,5-a]pyrimidine-6-carboxylic acid
CC1=NN2C(=C(C=NC2=C1)C(=O)O)C
This item is not returnable. View return policy
175201-51-1
C9H9N3O2
191.19
MFCD00067901
CACYYYIWDACOAQ-UHFFFAOYSA-N
2,7-dimethylpyrazolo 1,5-a pyrimidine-6-carboxylic acid, pyrazolo 1,5-a pyrimidine-6-carboxylic acid, 2,7-dimethyl, maybridge1_004672, 6-carboxy-2,7-dimethylpyrazolo 1,5-a pyrimidine, pyrazolo 1,5-a pyrimidine-6-carboxylicacid, 2,7-dimethyl, 2,7-dimethylpyrazolo 1,5-a pyrimidine-6-carboxylicacid, 2,7-dimethyl-8-hydropyrazolo 1,5-a pyrimidine-6-carboxylic acid
2736303
2,7-dimethylpyrazolo[1,5-a]pyrimidine-6-carboxylic acid
CC1=NN2C(=C(C=NC2=C1)C(=O)O)C
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 191.19
Percent Purity 97%
Chemical Name or Material 2, 7-Dimethylpyrazolo[1, 5-a]pyrimidine-6-carboxylic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'