Få mere at vide
2-[6-Methyl-2-(4-methylphenyl)imidazo[1,2-a]pyridin-3-yl]acetic Acid, Mikromol™
Artikelnummer. 17928034
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17928034 100 mg
17917284 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17928034 Leverandør Mikromol Leverandørnr. MM0333.03
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2-[6-Methyl-2-(4-methylphenyl)imidazo[1,2-a]pyridin-3-yl]acetic Acid
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-[6-Methyl-2-(4-methylphenyl)imidazo[1,2-a]pyridin-3-yl]acetic Acid
CAS 189005-44-5
Analyt- eller komponentnavne 2-[6-Methyl-2-(4-methylphenyl)imidazo[1,2-a]pyridin-3-yl]acetic Acid
Forsendelsestilstand Room Temperature
Molekylær formel C17 H16 N2 O2
Certificeringer/compliance ISO 17025
Synonym 2-[6-Methyl-2-(4-methylphenyl)imidazo[1,2-a]pyridin-3-yl]acetic Acid
InChI formel InChI=1S/C17H16N2O2/c1-11-3-6-13(7-4-11)17-14(9-16(20)21)19-10-12(2)5-8-15(19)18-17/h3-8,10H,9H2,1-2H3,(H,20,21)
SMIL Cc1ccc(cc1)c2nc3ccc(C)cn3c2CC(=O)O
IUPAC navn 2-[6-methyl-2-(4-methylphenyl)imidazo[1,2-a]pyridin-3-yl]acetic acid
Molekylvægt (g/mol) 280.32
Mængde 100 mg
Formel vægt 280.1212 g/mol